missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoropyridine-4-boronic acid, 98%

Manufacturer:  Alfa Aesar™ H53109MD

 View more versions of this product

Catalog No. AAH53109MD



Specifications

Specifications

3-Fluoropyridine-4-boronic acid
250mg
C5H5BFNO2
3-fluoropyridine-4-boronic acid, 3-fluoropyridin-4-yl boronic acid, 3-fluoropyridin-4-yl-4-boronic acid, 3-fluoro-4-pyridyl boronic acid, boronic acid, 3-fluoro-4-pyridinyl, 3-fluoro-4-pyridylboronic acid, 3-fluoropyridin-4-yl boranediol, 3-fluoro-4-pyridineboronic acid, 3-fluoropyridine-4-boronic acid, 3-fluoropyridine-4-boronicacid
B(C1=C(C=NC=C1)F)(O)O
140.908
140.91
237°C to 239°C
458532-97-3
MFCD03788558
QUHSESYLMZVXCN-UHFFFAOYSA-N
(3-fluoropyridin-4-yl)boronic acid
2779353
98%

Chemical Identifiers

458532-97-3
140.908
QUHSESYLMZVXCN-UHFFFAOYSA-N
2779353
B(C1=C(C=NC=C1)F)(O)O
C5H5BFNO2
MFCD03788558
3-fluoropyridine-4-boronic acid, 3-fluoropyridin-4-yl boronic acid, 3-fluoropyridin-4-yl-4-boronic acid, 3-fluoro-4-pyridyl boronic acid, boronic acid, 3-fluoro-4-pyridinyl, 3-fluoro-4-pyridylboronic acid, 3-fluoropyridin-4-yl boranediol, 3-fluoro-4-pyridineboronic acid, 3-fluoropyridine-4-boronic acid, 3-fluoropyridine-4-boronicacid
(3-fluoropyridin-4-yl)boronic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents