missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoropyridine-2-carboxaldehyde, 98%

Manufacturer:  Alfa Aesar™ H6190203

 View more versions of this product

Catalog No. AAH6190203



Description

Description

Applications
3-Fluoropyridine-2-carboxaldehyde is used to prepare taxoids derived from 9β-dihydrobaccatin-9,10-acetals with anti-tumor activities. It is also an intermediate used to synthesize S-3578 derivatives with potent activities against both methicillin-resistant Staphylococcus aureus (MRSA) and Pseudomonas aeruginosa.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

3-Fluoropyridine-2-carboxaldehyde
31224-43-8
MFCD07781234
Slightly soluble in water.
C1=CC(=C(N=C1)C=O)F
125.102
125.1
98%
1g
C6H4FNO
2-formyl-3-fluoropyridine
OZIMPUNGBUYCSP-UHFFFAOYSA-N
3-fluoropyridine-2-carbaldehyde
11344017
Solid

Chemical Identifiers

31224-43-8
125.102
OZIMPUNGBUYCSP-UHFFFAOYSA-N
11344017
C1=CC(=C(N=C1)C=O)F
C6H4FNO
MFCD07781234
2-formyl-3-fluoropyridine
3-fluoropyridine-2-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents