Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Fluoropropiophenone, 97%

$81.22 - $268.20

Chemical Identifiers

Chemical Name or Material 3'-Fluoropropiophenone
CAS 455-67-4
Molecular Formula C9H9FO
Formula Weight 152.17
MDL Number MFCD00061245
Beilstein 1936731
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1192203

  SDS
alfa aesar™
L1192203
1g Each for $81.22

AAL1192206

  SDS
alfa aesar™
L1192206
5g Each for $268.20
Description & Specifications

Specifications

Formula Weight 152.17
Health Hazard 1 Warning
Chemical Name or Material 3'-Fluoropropiophenone
Assay 97%
Molecular Formula C9H9FO
CAS 455-67-4
Beilstein 1936731
Quantity 1g
Boiling Point 94° to 96°C (5mmHg)
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
MDL Number MFCD00061245
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.