missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Fluoropropiophenone, 97%

Manufacturer:  Alfa Aesar™ L1192203

 View more versions of this product

Catalog No. AAL1192203



Specifications

Specifications

3'-Fluoropropiophenone
1g
C9H9FO
1936731
RPMOHVRRKYJFSB-UHFFFAOYSA-N
1-(3-fluorophenyl)propan-1-one
521183
97%
94°C to 96°C (5mmHg)
455-67-4
MFCD00061245
3'-fluoropropiophenone, 3-fluoropropiophenone, 1-3-fluorophenyl propan-1-one, 1-propanone, 1-3-fluorophenyl, 3-fluoro-propiophenone, 3;-fluoropropiophenone, 3-fluoro propiophenone, ethyl 3-fluorophenyl ketone, 1-3-fluorophenyl-1-propanone, 1-3-fluorophenyl-propan-1-one
CCC(=O)C1=CC(=CC=C1)F
152.168
152.17

Chemical Identifiers

455-67-4
152.168
RPMOHVRRKYJFSB-UHFFFAOYSA-N
521183
CCC(=O)C1=CC(=CC=C1)F
C9H9FO
MFCD00061245
3'-fluoropropiophenone, 3-fluoropropiophenone, 1-3-fluorophenyl propan-1-one, 1-propanone, 1-3-fluorophenyl, 3-fluoro-propiophenone, 3;-fluoropropiophenone, 3-fluoro propiophenone, ethyl 3-fluorophenyl ketone, 1-3-fluorophenyl-1-propanone, 1-3-fluorophenyl-propan-1-one
1-(3-fluorophenyl)propan-1-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents