missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Fluorophthalic Anhydride 98.0+%, TCI America™
Supplier: TCI America F034925G
Specifications
3-Fluorophthalic Anhydride | |
161°C | |
283°C | |
MFCD00039696 | |
3-fluorophthalic anhydride, 4-fluoroisobenzofuran-1,3-dione, 3-fluorophthalicanhydride, 4-fluoro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 4-fluoro, 5-fluoro-isobenzofurandione, 4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione, 4-fluor-2-benzofuran-1,3-dion, pubchem1949, fluorophthalic anhydride | |
FC1=CC=CC2=C1C(=O)OC2=O | |
166.11 | |
166.11 | |
Crystalline Powder |
652-39-1 | |
White-Yellow | |
C8H3FO3 | |
25 g | |
WWJAZKZLSDRAIV-UHFFFAOYSA-N | |
4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione | |
69551 | |
≥98.0% (GC) |
Chemical Identifiers
652-39-1 | |
166.11 | |
WWJAZKZLSDRAIV-UHFFFAOYSA-N | |
69551 | |
FC1=CC=CC2=C1C(=O)OC2=O |
C8H3FO3 | |
MFCD00039696 | |
3-fluorophthalic anhydride, 4-fluoroisobenzofuran-1,3-dione, 3-fluorophthalicanhydride, 4-fluoro-1,3-isobenzofurandione, 1,3-isobenzofurandione, 4-fluoro, 5-fluoro-isobenzofurandione, 4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione, 4-fluor-2-benzofuran-1,3-dion, pubchem1949, fluorophthalic anhydride | |
4-fluoro-1,3-dihydro-2-benzofuran-1,3-dione |
Safety and Handling
TSCA : No