Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluorophthalic acid, 97%

$36.80 - $464.86

Chemical Identifiers

Chemical Name or Material 3-Fluorophthalic acid
CAS 1583-67-1
Molecular Formula C8H5FO4
Formula Weight 184.12
MDL Number MFCD00039524
Beilstein 2616684
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1365603

 
alfa aesar™
A1365603
1g Each for $36.80

AAA1365606

 
alfa aesar™
A1365606
5g Each for $118.84

AAA1365614

 
alfa aesar™
A1365614
25g Each for $464.86
Description & Specifications

Specifications

Formula Weight 184.12
Chemical Name or Material 3-Fluorophthalic acid
Assay 97%
Molecular Formula C8H5FO4
MDL Number MFCD00039524
CAS 1583-67-1
Beilstein 2616684
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Odor Odorless
Melting Point ≈170°C (dec.)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.