Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluorophenylmagnesium bromide, 1M in MeTHF

Chemical Identifiers

Chemical Name or Material 3-Fluorophenylmagnesium bromide
CAS 17318-03-5
Molecular Formula C6H4BrFMg
Formula Weight 199.31
MDL Number MFCD00672007
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH51168AE

  SDS
alfa aesar™
H51168AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH51168AP

  SDS
alfa aesar™
H51168AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.31
CAS 17318-03-5
UN Number UN2924
Health Hazard 2 GHS H Statement
H225-H314-H302
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Concentration 1M in MeTHF
Quantity 100mL
Chemical Name or Material 3-Fluorophenylmagnesium bromide
Molecular Formula C6H4BrFMg
MDL Number MFCD00672007
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
Flash Point −11°C (12°F)

Applications
3-Fluorophenylmagnesium bromide is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF).

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents, acids, bases and alcohols.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H302
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if swallowed.