Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluorophenylglyoxal hydrate, 98%, dry wt. basis

$150.68 - $150.68

Chemical Identifiers

Chemical Name or Material 3-Fluorophenylglyoxal hydrate
CAS 121247-01-6
Molecular Formula C8H5FO2·xH2O
Formula Weight 152.13 (Anhydrous)
MDL Number MFCD05664101
Beilstein 3537863
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2027803

  SDS
alfa aesar™
L2027803
1g Each for $150.68
Description & Specifications

Specifications

Formula Weight 152.13 (Anhydrous)
CAS 121247-01-6
Chemical Name or Material 3-Fluorophenylglyoxal hydrate
MDL Number MFCD05664101
Beilstein 3537863
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Melting Point 66° to 68°C
Quantity 1g
Assay Percent Notes (dry wt.)
Molecular Formula C8H5FO2·xH2O
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.