Please login to your online account to display your discounted pricing

3-Fluorophenylacetone, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  350960050

 View more versions of this product

Catalog No. AC350960050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 152.17
Quantity 5g
Color Clear yellow-green
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 3-Fluorophenylacetone
Infrared Spectrum Authentic
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H9FO
MDL Number MFCD00061144
CAS 1737-19-5
CAS Min % 97.5
Packaging Glass bottle
Refractive Index 1.4955 to 1.4975 (20°C, 589nm)

Specifications

Chemical Name or Material 3-Fluorophenylacetone
Name Note 98%
CAS 1737-19-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H9FO
Formula Weight 152.17
MDL Number MFCD00061144