Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluorophenylacetone, 97%

$363.14 - $363.14

Chemical Identifiers

Chemical Name or Material 3-Fluorophenylacetone
CAS 1737-19-5
Molecular Formula C9H9FO
Formula Weight 152.17
MDL Number MFCD00061144
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2206106

  SDS
alfa aesar™
B2206106
5g Each for $363.14

AAB2206103

  SDS
alfa aesar™
B2206103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 3-Fluorophenylacetone
Quantity 5g
CAS 1737-19-5
Assay 97%
Molecular Formula C9H9FO
Formula Weight 152.17
MDL Number MFCD00061144

Applications
3-Fluorophenylacetone is used in the preparation of of optically active cyanohydrins. 3-Fluorophenylaceton is reactant in chiral amine synthesis.

Solubility
Soluble in chloroform, ethyl Acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.