Please login to your online account to display your discounted pricing

3-Fluorophenylacetic acid, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  278270250

 View more versions of this product

Catalog No. AC278270250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 154.14
Quantity 25g
Color White to pale yellow or light beige
Beilstein 09, 2261
EINECS Number 206-360-7
TSCA TSCA
Melting Point 44° to 48°C
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 97.5
Packaging Glass bottle
Chemical Name or Material 3-Fluorophenylacetic acid
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8 H7 F O2
Linear Formula FC6H4CH2COOH
MDL Number MFCD00004331
CAS 331-25-9

Specifications

Chemical Name or Material 3-Fluorophenylacetic acid
Name Note 98%
CAS 331-25-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8 H7 F O2
Linear Formula FC6H4CH2COOH
Formula Weight 154.14
MDL Number MFCD00004331