Please login to your online account to display your discounted pricing

3-Fluorophenylacetic acid, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Fluorophenylacetic acid
Name Note 98%
CAS 331-25-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8 H7 F O2
Linear Formula FC6H4CH2COOH
Formula Weight 154.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC278270250

  SDS
fisher scientific acros
278270250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 154.14
Quantity 25g
Color White to pale yellow or light beige
Beilstein 09, 2261
EINECS Number 206-360-7
TSCA TSCA
Melting Point 44° to 48°C
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 97.5
Packaging Glass bottle
Chemical Name or Material 3-Fluorophenylacetic acid
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8 H7 F O2
Linear Formula FC6H4CH2COOH
MDL Number MFCD00004331
CAS 331-25-9