Learn More
3-Fluorophenylacetic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1563506
Description
3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Fluorophenylacetic acid has been used as building block to synthesize the pentaamine and bis-heterocyclic libraries. It is also used as a medicine intermediates.
Solubility
Slightly soluble in chloroform and methanol. Insoluble in water.
Notes
Store at room temperature. Store in cool dry place in tightly closed container. Store away from oxidizing agent.
Specifications
3-Fluorophenylacetic acid | |
44°C to 48°C | |
>110°C (230°F) | |
C8H7FO2 | |
5 g | |
3-fluorophenylacetic acid, 2-3-fluorophenyl acetic acid, m-fluorophenylacetic acid, benzeneacetic acid, 3-fluoro, 3-fluorophenyl acetic acid, 3-fluorophenlacetic acid, acetic acid, m-fluorophenyl, m-fluorophenyl acetic acid, 3-fluorophenylaceticacid | |
YEAUYVGUXSZCFI-UHFFFAOYSA-N | |
2-(3-fluorophenyl)acetic acid | |
67617 | |
98% |
331-25-9 | |
151°C (0.8 mmHg) | |
Odorless | |
MFCD00004331 | |
775898 | |
Slightly soluble in chloroform and methanol. Insoluble in water. | |
OC(=O)CC1=CC=CC(F)=C1 | |
154.14 | |
154.14 |
Chemical Identifiers
331-25-9 | |
154.14 | |
YEAUYVGUXSZCFI-UHFFFAOYSA-N | |
67617 | |
OC(=O)CC1=CC=CC(F)=C1 |
C8H7FO2 | |
MFCD00004331 | |
3-fluorophenylacetic acid, 2-3-fluorophenyl acetic acid, m-fluorophenylacetic acid, benzeneacetic acid, 3-fluoro, 3-fluorophenyl acetic acid, 3-fluorophenlacetic acid, acetic acid, m-fluorophenyl, m-fluorophenyl acetic acid, 3-fluorophenylaceticacid | |
2-(3-fluorophenyl)acetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 206-360-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only