Learn More
3-Fluorophenyl isocyanate, 97+%, Thermo Scientific Chemicals
$115.63 - $393.63
Chemical Identifiers
CAS | 404-71-7 |
---|---|
Molecular Formula | C7H4FNO |
Molecular Weight (g/mol) | 137.11 |
MDL Number | MFCD00002015 |
InChI Key | RIKWVZGZRYDACA-UHFFFAOYSA-N |
Synonym | 3-fluorophenyl isocyanate, m-fluorophenyl isocyanate, 3-fluorophenylisocyanate, m-fluorophenylisocyanate, benzene, 1-fluoro-3-isocyanato, 3-isocyanatofluorobenzene, isocyanic acid 3-fluorophenyl ester, 3-fluorobenzenisocyanate, pubchem4995, acmc-1ajez |
PubChem CID | 123064 |
IUPAC Name | 1-fluoro-3-isocyanatobenzene |
SMILES | FC1=CC=CC(=C1)N=C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0149606
|
Thermo Scientific Chemicals
L0149606 |
5 g |
Each for $115.63
|
|
|||||
AAL0149614
|
Thermo Scientific Chemicals
L0149614 |
25 g |
Each for $393.63
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
404-71-7 | |
137.11 | |
RIKWVZGZRYDACA-UHFFFAOYSA-N | |
123064 | |
FC1=CC=CC(=C1)N=C=O |
C7H4FNO | |
MFCD00002015 | |
3-fluorophenyl isocyanate, m-fluorophenyl isocyanate, 3-fluorophenylisocyanate, m-fluorophenylisocyanate, benzene, 1-fluoro-3-isocyanato, 3-isocyanatofluorobenzene, isocyanic acid 3-fluorophenyl ester, 3-fluorobenzenisocyanate, pubchem4995, acmc-1ajez | |
1-fluoro-3-isocyanatobenzene |
Specifications
404-71-7 | |
56°C to 58°C (14 mmHg) | |
Pungent, Makes Eyes Water | |
1.5141 | |
5 g | |
774879 | |
3-fluorophenyl isocyanate, m-fluorophenyl isocyanate, 3-fluorophenylisocyanate, m-fluorophenylisocyanate, benzene, 1-fluoro-3-isocyanato, 3-isocyanatofluorobenzene, isocyanic acid 3-fluorophenyl ester, 3-fluorobenzenisocyanate, pubchem4995, acmc-1ajez | |
FC1=CC=CC(=C1)N=C=O | |
137.11 | |
137.11 | |
3-Fluorophenyl isocyanate |
1.207 | |
41°C (105°F) | |
C7H4FNO | |
MFCD00002015 | |
UN3080 | |
Moisture sensitive | |
RIKWVZGZRYDACA-UHFFFAOYSA-N | |
1-fluoro-3-isocyanatobenzene | |
123064 | |
≥97% |
Safety and Handling
GHS H Statement
H300-H330-H334-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P285-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H301+H311+H331-H315-H319-H334-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
EINECSNumber : 206-966-1
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only