Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluorophenyl acetate, 98%

$57.57 - $57.57

Chemical Identifiers

Chemical Name or Material 3-Fluorophenyl acetate
CAS 701-83-7
Molecular Formula C8H7FO2
Formula Weight 154.14
MDL Number MFCD00142578
Beilstein 2555261
Synonym 1-Acetoxy-3-fluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1346506

  SDS
alfa aesar™
L1346506
5g Each for $57.57
Description & Specifications

Specifications

Formula Weight 154.14
Molecular Formula C8H7FO2
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Fluorophenyl acetate
Assay 98%
MDL Number MFCD00142578
CAS 701-83-7
Beilstein 2555261
Synonym 1-Acetoxy-3-fluorobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Flash Point 76°C (169°F)
Quantity 5g
Boiling Point 77° to 78°C (13mmHg)
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.