missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluorophenyl acetate, 98%

Manufacturer:  Alfa Aesar™ L1346506

 View more versions of this product

Catalog No. AAL1346506



Specifications

Specifications

3-Fluorophenyl acetate
77°C to 78°C (13mmHg)
701-83-7
MFCD00142578
1-acetoxy-3-fluorobenzene, 3-fluorophenylacetate, acetic acid 3-fluorophenyl ester, pubchem3060, m-fluorophenyl acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-, acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-,1-acetate, iawzwmgutkrlqb-uhfffaoysa
CC(=O)OC1=CC(=CC=C1)F
154.14
154.14
76°C (169°F)
5g
C8H7FO2
2555261
IAWZWMGUTKRLQB-UHFFFAOYSA-N
(3-fluorophenyl) acetate
136542
98%

Chemical Identifiers

701-83-7
154.14
IAWZWMGUTKRLQB-UHFFFAOYSA-N
136542
CC(=O)OC1=CC(=CC=C1)F
C8H7FO2
MFCD00142578
1-acetoxy-3-fluorobenzene, 3-fluorophenylacetate, acetic acid 3-fluorophenyl ester, pubchem3060, m-fluorophenyl acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-, acetate, 3-fluorophenyl acetate, phenol, 3-fluoro-,1-acetate, iawzwmgutkrlqb-uhfffaoysa
(3-fluorophenyl) acetate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents