missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluorophenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 119550100

 View more versions of this product

Catalog No. AC119550100



Specifications

Specifications

3-Fluorophenol
71°C
Glass bottle
1.24
Brown-Yellow to Colorless
10mL
97.5
97.5% min. (GC)
FC6H4OH
06, I, 97
Solubility in water: insoluble
C1=CC(=CC(=C1)F)O
112.1
112.1
98%
1.2400g/mL
Authentic
1.5125 to 1.5155
178.0°C
10.0°C
372-20-3
100.0
C6H5FO
MFCD00002254
3-fluoro phenol, 3-fluoro-phenol, 3-fluorophenol, 3-fluorphenol, m-fluoro-phenol, m-fluorophenol, meta-fluorophenol, phenol, 3-fluoro, phenol, m-fluoro, unii-t7oma38487
SJTBRFHBXDZMPS-UHFFFAOYSA-N
3-fluorophenol
9743
Liquid

Chemical Identifiers

372-20-3
112.1
SJTBRFHBXDZMPS-UHFFFAOYSA-N
9743
C1=CC(=CC(=C1)F)O
C6H5FO
MFCD00002254
3-fluoro phenol, 3-fluoro-phenol, 3-fluorophenol, 3-fluorphenol, m-fluoro-phenol, m-fluorophenol, meta-fluorophenol, phenol, 3-fluoro, phenol, m-fluoro, unii-t7oma38487
3-fluorophenol
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 206-748-6

Recommended Storage : this product darkens during storage

SDS Documents