missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluorophenethylamine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC350941000

 View more versions of this product

Catalog No. AC350941000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
139.17
Clear colorless to pale yellow
100.0
3-Fluorophenethylamine
1.5080 to 1.5100 (20°C, 589nm)
C8H10FN
MFCD00075376
Authentic
Liquid
100g
1.0660g/mL
Glass bottle
FC6H4CHCHNH
404-70-6
99%
98.5

Chemical Identifiers

404-70-6
100.0
C8H10FN
139.17
3-Fluorophenethylamine
98.5
99%
FC6H4CHCHNH
MFCD00075376
≥98.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger