Please login to your online account to display your discounted pricing

3'-Fluorobenzylspiperone maleate, Tocris


D2-like receptor ligand

Quantity 10mg

Manufacturer: r & d systems  070110

 View more versions of this product

Catalog No. 070110

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 619.67
CAS 1135278-61-3
Recommended Storage Store at Room Temperature
Chemical Name or Material 3'-Fluorobenzylspiperone maleate
Synonym 8-[4-(4-Fluorophenyl)-4-oxobutyl]-(3-fluorobenzyl-1-phenyl)-1,3,8-triazaspiro[4,5]decan-4-one maleate
Molecular Formula C30H31F2N3O2·C4H4O4
Assay Percent Range >98%

Specifications

Chemical Name or Material 3'-Fluorobenzylspiperone maleate
Assay Percent Range >98%
Molecular Formula C30H31F2N3O2·C4H4O4
Formula Weight 619.67
Synonym 8-[4-(4-Fluorophenyl)-4-oxobutyl]-(3-fluorobenzyl-1-phenyl)-1,3,8-triazaspiro[4,5]decan-4-one maleate
Accessories

Store at Room Temperature