Please login to your online account to display your discounted pricing

3'-Fluorobenzylspiperone maleate, Tocris

D2-like receptor ligand

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material 3'-Fluorobenzylspiperone maleate
Assay Percent Range >98%
Molecular Formula C30H31F2N3O2·C4H4O4
Formula Weight 619.67
Synonym 8-[4-(4-Fluorophenyl)-4-oxobutyl]-(3-fluorobenzyl-1-phenyl)-1,3,8-triazaspiro[4,5]decan-4-one maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

070110

 
r & d systems
070110
10mg Each for $164.61

070150

 
r & d systems
070150
50mg Each for $682.67
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 619.67
CAS 1135278-61-3
Recommended Storage Store at Room Temperature
Chemical Name or Material 3'-Fluorobenzylspiperone maleate
Synonym 8-[4-(4-Fluorophenyl)-4-oxobutyl]-(3-fluorobenzyl-1-phenyl)-1,3,8-triazaspiro[4,5]decan-4-one maleate
Molecular Formula C30H31F2N3O2·C4H4O4
Assay Percent Range >98%
Safety and Handling

Store at Room Temperature