Please login to your online account to display your discounted pricing

3-Fluorobenzyl Chloride 96%, ACROS Organics™


Quantity 50mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  119420500

 View more versions of this product

Catalog No. AC119420500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid
Formula Weight 144.58
Quantity 50mL
Color Clear colorless to light yellow
EINECS Number 207-264-8
TSCA TSCA
Density 1.2100g/mL
CAS Min % 95.0
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym α-Chloro-m-fluorotoluene
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.5115 to 1.5135 (20°C, 589nm)
Chemical Name or Material 3-Fluorobenzyl chloride
Name Note 96%
Assay Percent Range 96%
Molecular Formula C7H6ClF
Linear Formula FC6H4CH2Cl
MDL Number MFCD00000904
CAS 456-42-8
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

Specifications

Chemical Name or Material 3-Fluorobenzyl chloride
Name Note 96%
CAS 456-42-8
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H6ClF
Linear Formula FC6H4CH2Cl
Formula Weight 144.58
MDL Number MFCD00000904
Synonym α-Chloro-m-fluorotoluene
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

SDS & Safety Documents