missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluorobenzyl Chloride 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 119420100

 View more versions of this product

Catalog No. AC119420100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥95%
144.58
Clear colorless to light yellow
100.0
1.5115 to 1.5135 (20°C, 589nm)
α-Chloro-m-fluorotoluene
FC6H4CH2Cl
96%
96%
C7H6ClF
Liquid
10mL
1.2100g/mL
95.0
Authentic
Glass bottle
MFCD00000904
456-42-8
3-Fluorobenzyl chloride

Chemical Identifiers

456-42-8
100.0
C7H6ClF
144.58
α-Chloro-m-fluorotoluene
≥95%
95.0
96%
FC6H4CH2Cl
MFCD00000904
3-Fluorobenzyl chloride, 96%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-264-8

TSCA : TSCA