missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluorobenzyl Bromide, TCI America™

Manufacturer:  TCI America F0208250G

 View more versions of this product

Catalog No. F0208250G



Specifications

Specifications

3-Fluorobenzyl Bromide
Yellow
456-41-7
MFCD00000340
3-fluorobenzyl bromide, 1-bromomethyl-3-fluorobenzene, 3-fluorobenzylbromide, m-fluorobenzyl bromide, benzene, 1-bromomethyl-3-fluoro, m-fluorobenzylbromide, alpha-bromo-m-fluorotoluene, alpha-bromo-3-fluorotoluene, toluene, .alpha.-bromo-m-fluoro, 1-bromobenzyl-3-fluorobenzene
C1=CC(=CC(=C1)F)CBr
189.027
189.03
≥97.0% (GC)
200°C
250g
C7H6BrF
3265
SCBZBMXPJYMXRC-UHFFFAOYSA-N
1-(bromomethyl)-3-fluorobenzene
68007
Clear Liquid at 20°C

Chemical Identifiers

456-41-7
189.027
SCBZBMXPJYMXRC-UHFFFAOYSA-N
68007
C1=CC(=CC(=C1)F)CBr
C7H6BrF
MFCD00000340
3-fluorobenzyl bromide, 1-bromomethyl-3-fluorobenzene, 3-fluorobenzylbromide, m-fluorobenzyl bromide, benzene, 1-bromomethyl-3-fluoro, m-fluorobenzylbromide, alpha-bromo-m-fluorotoluene, alpha-bromo-3-fluorotoluene, toluene, .alpha.-bromo-m-fluoro, 1-bromobenzyl-3-fluorobenzene
1-(bromomethyl)-3-fluorobenzene
Safety and Handling

Safety and Handling

TSCA : Yes

Documents