Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluorobenzyl bromide, 98%

Chemical Identifiers

Chemical Name or Material 3-Fluorobenzyl bromide
CAS 456-41-7
Molecular Formula C7H6BrF
Formula Weight 189.03
MDL Number MFCD00000340
UN Number UN3265
Beilstein 636503
Synonym alpha-Bromo-3-fluorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1544606

 
alfa aesar™
A1544606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1544614

 
alfa aesar™
A1544614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1544618

 
alfa aesar™
A1544618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 189.03
Chemical Name or Material 3-Fluorobenzyl bromide
TSCA TSCA
Assay 98%
Molecular Formula C7H6BrF
MDL Number MFCD00000340
CAS 456-41-7
UN Number UN3265
Beilstein 636503
Synonym alpha-Bromo-3-fluorotoluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Flash Point 91°C (196°F)
Odor Pungent
Boiling Point 200°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.