missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluorobenzyl bromide, 98%

Manufacturer:  Alfa Aesar™ A1544618

 View more versions of this product

Catalog No. AAA1544618



Specifications

Specifications

3-Fluorobenzyl bromide
Pungent
50g
C7H6BrF
UN3265
3-fluorobenzyl bromide, 1-bromomethyl-3-fluorobenzene, 3-fluorobenzylbromide, m-fluorobenzyl bromide, benzene, 1-bromomethyl-3-fluoro, m-fluorobenzylbromide, alpha-bromo-m-fluorotoluene, alpha-bromo-3-fluorotoluene, toluene, .alpha.-bromo-m-fluoro, 1-bromobenzyl-3-fluorobenzene
C1=CC(=CC(=C1)F)CBr
189.027
189.03
91°C (196°F)
200°C
456-41-7
MFCD00000340
636503
SCBZBMXPJYMXRC-UHFFFAOYSA-N
1-(bromomethyl)-3-fluorobenzene
68007
98%

Chemical Identifiers

456-41-7
189.027
SCBZBMXPJYMXRC-UHFFFAOYSA-N
68007
C1=CC(=CC(=C1)F)CBr
C7H6BrF
MFCD00000340
3-fluorobenzyl bromide, 1-bromomethyl-3-fluorobenzene, 3-fluorobenzylbromide, m-fluorobenzyl bromide, benzene, 1-bromomethyl-3-fluoro, m-fluorobenzylbromide, alpha-bromo-m-fluorotoluene, alpha-bromo-3-fluorotoluene, toluene, .alpha.-bromo-m-fluoro, 1-bromobenzyl-3-fluorobenzene
1-(bromomethyl)-3-fluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Documents