Please login to your online account to display your discounted pricing

3-Fluorobenzoyl chloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Fluorobenzoyl chloride
CAS 1711-07-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4ClFO
Linear Formula FC6H4COCl
Formula Weight 158.56
MDL Number MFCD00000670
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC171981000

  SDS
thermo fisher scientific
AC171981000
Glass bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC171980250

  SDS
thermo fisher scientific
171980250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC171980050

  SDS
thermo fisher scientific
171980050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 158.56
Physical Form Liquid
Quantity 100mL
Color Clear colorless to light yellow
Density 1.3000g/mL
Beilstein 09, I, 137
CAS Min % 96.0
CAS Max % 100.0
Linear Formula FC6H4COCl
CAS 1711-07-5
Infrared Spectrum Authentic
Refractive Index 1.5260 to 1.5300 (20°C, 589nm)
Chemical Name or Material 3-Fluorobenzoyl chloride
Assay Percent Range 97%
Molecular Formula C7H4ClFO
MDL Number MFCD00000670
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.