Please login to your online account to display your discounted pricing

3-Fluorobenzoyl chloride, 97%, ACROS Organics™


Quantity 25mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  171980250

 View more versions of this product

Catalog No. AC171980250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 158.56
Quantity 25mL
Color Clear colorless to light yellow
Beilstein 09, I, 137
Density 1.3000g/mL
CAS Max % 100.0
CAS Min % 96.0
Infrared Spectrum Authentic
Chemical Name or Material 3-Fluorobenzoyl chloride
Assay Percent Range 97%
CAS 1711-07-5
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00000670
Packaging Glass bottle
Refractive Index 1.5260 to 1.5300 (20°C, 589nm)
Linear Formula FC6H4COCl
Molecular Formula C7H4ClFO
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 3-Fluorobenzoyl chloride
CAS 1711-07-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H4ClFO
Linear Formula FC6H4COCl
Formula Weight 158.56
MDL Number MFCD00000670
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

SDS & Safety Documents