missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluorobenzoic hydrazide, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC436340010

 View more versions of this product

Catalog No. AC436340010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

3-Fluorobenzoic hydrazide
Glass Bottle
140.0°C to 141.0°C
499-55-8
100.0
C7H7FN2O
UUISEXNUHLZEDB-UHFFFAOYSA-N
3-fluorobenzohydrazide
351640
Crystalline Powder
Authentic
White
1g
96.0
96% min. (HPLC)
#, 1-fluorobenzene-3-carbohydrazide, 3-fluoro-benzoic acid hydrazide, 3-fluorobenzenecarbohydrazide, 3-fluorobenzhydrazide, 3-fluorobenzoic acid hydrazide, 3-fluorobenzoic hydrazide, acmc-20an3d, ksc497s6p, pubchem3482
C1=CC(=CC(=C1)F)C(=O)NN
154.15
154.15
97%

Chemical Identifiers

499-55-8
154.15
#, 1-fluorobenzene-3-carbohydrazide, 3-fluoro-benzoic acid hydrazide, 3-fluorobenzenecarbohydrazide, 3-fluorobenzhydrazide, 3-fluorobenzoic acid hydrazide, 3-fluorobenzoic hydrazide, acmc-20an3d, ksc497s6p, pubchem3482
3-fluorobenzohydrazide
C7H7FN2O
UUISEXNUHLZEDB-UHFFFAOYSA-N
351640
C1=CC(=CC(=C1)F)C(=O)NN
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

SDS Documents