missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluorobenzenesulfonyl chloride, 98%, ACROS Organics™

Chemical Identifiers

CAS 701-27-9
Molecular Formula C6H4ClFO2S
Molecular Weight (g/mol) 194.61
MDL Number MFCD00042286
InChI Key OKYSUJVCDXZGKE-UHFFFAOYSA-N
Synonym 3-fluoro-benzenesulfonyl chloride
PubChem CID 2734258
IUPAC Name 3-fluorobenzenesulfonyl chloride
SMILES C1=CC(=CC(=C1)S(=O)(=O)Cl)F
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC356930010 SDS View Documents Thermo Fisher Scientific
AC356930010
1g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

701-27-9
194.61
OKYSUJVCDXZGKE-UHFFFAOYSA-N
2734258
C1=CC(=CC(=C1)S(=O)(=O)Cl)F
C6H4ClFO2S
MFCD00042286
3-fluoro-benzenesulfonyl chloride
3-fluorobenzenesulfonyl chloride

Specifications

Authentic
1.5280 to 1.5310
Brown to Brown-Gray
701-27-9
100.0
C6H4ClFO2S
MFCD00042286
OKYSUJVCDXZGKE-UHFFFAOYSA-N
3-fluorobenzenesulfonyl chloride
2734258
Powder, Granules or Chunks
3-Fluorobenzenesulfonyl chloride, 98%
Glass bottle
108.0°C (15.0 mmHg)
1g
97.5
97.5% min. (GC)
FC6H4SO2Cl
3-fluoro-benzenesulfonyl chloride
C1=CC(=CC(=C1)S(=O)(=O)Cl)F
194.61
194.61
98%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger