missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluorobenzaldehyde, 97%

Manufacturer:  Alfa Aesar™ A1139618

 View more versions of this product

Catalog No. AAA1139618



Specifications

Specifications

3-Fluorobenzaldehyde
173°C
456-48-4
MFCD00003348
970178
PIKNVEVCWAAOMJ-UHFFFAOYSA-N
3-fluorobenzaldehyde
68009
97%
56°C (132°F)
50g
C7H5FO
UN1989
m-fluorobenzaldehyde, benzaldehyde, 3-fluoro, 3'-fluoro-benzaldehyde, 3-fluoro-benzaldehyde, meta-fluorobenzaldehyde, benzaldehyde, m-fluoro, 3-fc6h4cho, 3-fluorobenzaldehyde, 3-fluoro benzaldehyde, pubchem3477
C1=CC(=CC(=C1)F)C=O
124.114
124.11

Chemical Identifiers

456-48-4
124.114
PIKNVEVCWAAOMJ-UHFFFAOYSA-N
68009
C1=CC(=CC(=C1)F)C=O
C7H5FO
MFCD00003348
m-fluorobenzaldehyde, benzaldehyde, 3-fluoro, 3'-fluoro-benzaldehyde, 3-fluoro-benzaldehyde, meta-fluorobenzaldehyde, benzaldehyde, m-fluoro, 3-fc6h4cho, 3-fluorobenzaldehyde, 3-fluoro benzaldehyde, pubchem3477
3-fluorobenzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALDEHYDES, N.O.S.

TSCA : TSCA

SDS Documents