Learn More
3-Fluoroaniline, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1422814
Description
3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Fluoroaniline is used in the preparation of acetic acid-(3-fluoro-anilide by reacting with acetic acid anhydride. Further, it serves as a reagent in the preparation of radamide analogs as Grp94 inhibitors, which has potential to be antitumor agents.
Solubility
Miscible with ethanol and ethyl ether. Slightly miscible with chloroform and water.
Notes
Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
Specifications
3-Fluoroaniline | |
1.156 | |
77°C (170°F) | |
C6H6FN | |
MFCD00007758 | |
UN2941 | |
m-fluoroaniline, 1-amino-3-fluorobenzene, 3-fluoranilin, benzenamine, 3-fluoro, 3-fluorobenzenamine, 3-fluoro-phenylamine, aniline, 3-fluoro, 3-fluorophenylamine, aniline, m-fluoro, 3-fluoranilin czech | |
QZVQQUVWFIZUBQ-UHFFFAOYSA-N | |
3-fluoroaniline | |
9742 | |
111.12 |
372-19-0 | |
186°C to 187°C | |
Recognizable | |
1.544 | |
25 g | |
1305471 | |
Miscible with ethanol and ethyl ether. Slightly miscible with chloroform and water. | |
C1=CC(=CC(=C1)F)N | |
111.119 | |
CHEBI:27873 | |
≥98% |
Chemical Identifiers
372-19-0 | |
111.119 | |
QZVQQUVWFIZUBQ-UHFFFAOYSA-N | |
9742 | |
3-fluoroaniline |
C6H6FN | |
MFCD00007758 | |
m-fluoroaniline, 1-amino-3-fluorobenzene, 3-fluoranilin, benzenamine, 3-fluoro, 3-fluorobenzenamine, 3-fluoro-phenylamine, aniline, 3-fluoro, 3-fluorophenylamine, aniline, m-fluoro, 3-fluoranilin czech | |
CHEBI:27873 | |
C1=CC(=CC(=C1)F)N |
Safety and Handling
GHS H Statement
H331-H373-H227-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H318-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FLUOROANILINES
EINECSNumber : 206-747-0
RTECSNumber : BY1400000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only