missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3′-Fluoroacetophenone, TCI America™

Manufacturer:  TCI America F026225G

 View more versions of this product

Catalog No. F026225G



Specifications

Specifications

3′-Fluoroacetophenone
Yellow
25g
C8H7FO
3'-fluoroacetophenone, 1-3-fluorophenyl ethanone, 3-fluoroacetophenone, m-fluoroacetophenone, ethanone, 1-3-fluorophenyl, 1-3-fluorophenyl ethan-1-one, acetophenone, 3'-fluoro, 3'-fluoro-acetophenone, 1-acetyl-3-fluorobenzene, ethanone, 1-3-fluorophenyl
CC(=O)C1=CC(=CC=C1)F
138.141
138.14
≥97.0% (GC)
81°C
-3°C
455-36-7
MFCD00000338
HCEKGPAHZCYRBZ-UHFFFAOYSA-N
1-(3-fluorophenyl)ethanone
9967
Clear Liquid at 20°C

Chemical Identifiers

455-36-7
138.141
HCEKGPAHZCYRBZ-UHFFFAOYSA-N
9967
CC(=O)C1=CC(=CC=C1)F
C8H7FO
MFCD00000338
3'-fluoroacetophenone, 1-3-fluorophenyl ethanone, 3-fluoroacetophenone, m-fluoroacetophenone, ethanone, 1-3-fluorophenyl, 1-3-fluorophenyl ethan-1-one, acetophenone, 3'-fluoro, 3'-fluoro-acetophenone, 1-acetyl-3-fluorobenzene, ethanone, 1-3-fluorophenyl
1-(3-fluorophenyl)ethanone
Safety and Handling

Safety and Handling

TSCA : No

Documents