Please login to your online account to display your discounted pricing

3'-Fluoroacetophenone, 99%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC204440250

 View more versions of this product

Catalog No. AC204440250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 138.14
Quantity 25g
Color Clear colorless to yellow
Density 1.1200g/mL
Beilstein 07, III, 961
CAS Max % 100.0
Chemical Name or Material 3';-Fluoroacetophenone
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C8H7FO
MDL Number MFCD00000338
CAS 455-36-7
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Refractive Index 1.5080 to 1.5100 (20°C, 589nm)
Linear Formula FC6H4COCH3
Synonym 1-(3-Fluorophenyl)ethanone
CAS Min % 98.5
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Specifications

Chemical Name or Material 3';-Fluoroacetophenone
CAS 455-36-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H7FO
Linear Formula FC6H4COCH3
Formula Weight 138.14
MDL Number MFCD00000338
Synonym 1-(3-Fluorophenyl)ethanone
Accessories

Warning

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS P Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Causes skin irritation.

SDS & Safety Documents