missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3'-Fluoroacetophenone, 99%, ACROS Organics™

Chemical Identifiers

CAS 455-36-7
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Molecular Formula C8H7FO
Linear Formula FC6H4COCH3
Formula Weight 138.14
MDL Number MFCD00000338
Synonym 1-(3-Fluorophenyl)ethanone
Chemical Name or Material 3';-Fluoroacetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC204440250 SDS Thermo Fisher Scientific
AC204440250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC204440010 SDS Thermo Fisher Scientific
204440010
Glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC204440050 SDS Fisher Scientific Acros
204440050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

455-36-7
100.0
C8H7FO
138.14
1-(3-Fluorophenyl)ethanone
≥98.5%
98.5
99%
FC6H4COCH3
MFCD00000338
3';-Fluoroacetophenone

Specifications

Clear colorless to yellow
455-36-7
100.0
Authentic
1.5080 to 1.5100 (20°C, 589nm)
99%
FC6H4COCH3
MFCD00000338
1-(3-Fluorophenyl)ethanone
≥98.5%
25g
98.5
1.1200g/mL
Glass bottle
Liquid
C8H7FO
138.14
07, III, 961
3';-Fluoroacetophenone

Safety and Handling

GHS P Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Causes skin irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning