Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Fluoroacetophenone, 98+%

$81.12 - $1,092.89

Chemical Identifiers

Chemical Name or Material 3'-Fluoroacetophenone
CAS 455-36-7
Molecular Formula C8H7FO
Formula Weight 138.14
MDL Number MFCD00000338
Beilstein 907041
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1173306

 
alfa aesar™
A1173306
5g Each for $81.12

AAA1173322

 
alfa aesar™
A1173322
100g Each for $1,092.89
Description & Specifications

Specifications

Formula Weight 138.14
Chemical Name or Material 3'-Fluoroacetophenone
Assay ≥98%
Molecular Formula C8H7FO
MDL Number MFCD00000338
CAS 455-36-7
Beilstein 907041
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 80°C (176°F)
Boiling Point 80° to 81°C (9mmHg)
Melting Point −3°C
Quantity 5g

Applications
3′-Fluoroacetophenone is used as raw material and intermediate in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents. Ensure adequate ventilation during use. Use only in a chemical fume hood.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.