Please login to your online account to display your discounted pricing

Alfa Aesar™ 3'-Fluoroacetanilide, 98%

$34.66 - $326.97

Chemical Identifiers

Chemical Name or Material 3'-Fluoroacetanilide
CAS 351-28-0
Molecular Formula C8H8FNO
Formula Weight 153.16
MDL Number MFCD00017917
Beilstein 2803073
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2288906

  SDS
alfa aesar™
B2288906
5g Each for $34.66

AAB2288914

  SDS
alfa aesar™
B2288914
25g Each for $110.30

AAB2288922

  SDS
alfa aesar™
B2288922
100g Each for $326.97
Description & Specifications

Specifications

Formula Weight 153.16
Molecular Formula C8H8FNO
Melting Point 86° to 89°C
Beilstein 2803073
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3'-Fluoroacetanilide
MDL Number MFCD00017917
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
CAS 351-28-0
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.