Learn More
3-Fluoro-o-xylene, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1063714
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.
Solubility
Insoluble in water.
Notes
Stable under normal conditions. Incompatible with oxidizing agents.
Specifications
3-Fluoro-o-xylene | |
0.992 | |
25 g | |
1.486 | |
MFCD00000323 | |
2040955 | |
Insoluble in water. | |
CC1=CC=CC(F)=C1C | |
124.16 | |
124.16 |
443-82-3 | |
147°C to 148°C | |
Aromatic | |
C8H9F | |
UN1993 | |
3-fluoro-o-xylene, 2,3-dimethylfluorobenzene, 3-fluoro-1,2-xylene, o-xylene, 3-fluoro, 3-fluoro-1,2-dimethylbenzene, benzene, 1-fluoro-2,3-dimethyl, 1,2-dimethyl-3-fluorobenzene, 1-fluoro-2,3-dimethyl-benzene, fluorxylol, pubchem4408 | |
AWLDSXJCQWTJPC-UHFFFAOYSA-N | |
1-fluoro-2,3-dimethylbenzene | |
96489 | |
99% |
Chemical Identifiers
443-82-3 | |
124.16 | |
AWLDSXJCQWTJPC-UHFFFAOYSA-N | |
96489 | |
CC1=CC=CC(F)=C1C |
C8H9F | |
MFCD00000323 | |
3-fluoro-o-xylene, 2,3-dimethylfluorobenzene, 3-fluoro-1,2-xylene, o-xylene, 3-fluoro, 3-fluoro-1,2-dimethylbenzene, benzene, 1-fluoro-2,3-dimethyl, 1,2-dimethyl-3-fluorobenzene, 1-fluoro-2,3-dimethyl-benzene, fluorxylol, pubchem4408 | |
1-fluoro-2,3-dimethylbenzene |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c
H226-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 207-140-3
RTECSNumber : ZE4560000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only