missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-N-methylbenzylamine, 97%

Manufacturer:  Alfa Aesar™ H5617706

 View more versions of this product

Catalog No. AAH5617706



Specifications

Specifications

3-Fluoro-N-methylbenzylamine
66°C (151°F)
183°C to 184°C
5g
C8H10FN
1-3-fluorophenyl-n-methyl-methanamine
CNCC1=CC(=CC=C1)F
139.173
139.17
97%
1.015 g/mL
1.499
Colorless to Tan
90389-84-7
MFCD04623547
ZXWCKKSSCIFVBT-UHFFFAOYSA-N
1-(3-fluorophenyl)-N-methylmethanamine
457577
Liquid

Chemical Identifiers

90389-84-7
139.173
ZXWCKKSSCIFVBT-UHFFFAOYSA-N
457577
CNCC1=CC(=CC=C1)F
C8H10FN
MFCD04623547
1-3-fluorophenyl-n-methyl-methanamine
1-(3-fluorophenyl)-N-methylmethanamine
Safety and Handling

Safety and Handling

P210-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P330-P370+P378q-P501c

H227-H302-H318

TSCA : No

Recommended Storage : Ambient temperatures

Documents