Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-N-methylbenzylamine, 97%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-N-methylbenzylamine
CAS 90389-84-7
Formula Weight 139.17
MDL Number MFCD04623547
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5617706

 
alfa aesar™
H5617706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5617714

 
alfa aesar™
H5617714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 139.17
Boiling Point 183° to 184°C
Assay 97%
MDL Number MFCD04623547
CAS 90389-84-7
Color Colorless to tan
Quantity 5g
Chemical Name or Material 3-Fluoro-N-methylbenzylamine
Physical Form Liquid
Refractive Index 1.499
Density 1.015g/mL