Learn More
3-Fluoro-DL-tyrosine, 97%, Thermo Scientific Chemicals
$136.36 - $535.29
Chemical Identifiers
CAS | 403-90-7 |
---|---|
Molecular Formula | C9H10FNO3 |
Molecular Weight (g/mol) | 199.18 |
MDL Number | MFCD00063075 |
InChI Key | VIIAUOZUUGXERI-UHFFFAOYNA-N |
Synonym | 3-fluoro-dl-tyrosine, 2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, pardinon, m-fluoro-dl-tyrosine, fluorthyrin, 3-fluorotyrosin, 3-fluorotyrosine, m-ftyr, dl-m-fluorotyrosine, dl-3-fluorotyrosine |
PubChem CID | 92100 |
ChEBI | CHEBI:32771 |
IUPAC Name | 2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid |
SMILES | NC(CC1=CC=C(O)C(F)=C1)C(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL01479MC
|
Thermo Scientific Chemicals
L01479MC |
100 mg |
Each for $136.36
|
|
|||||
AAL01479ME
|
Thermo Scientific Chemicals
L01479ME |
500 mg |
Each for $535.29
|
|
|||||
Description
3-Fluoro-DL-tyrosine is used for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Fluoro-DL-tyrosine is used for tyrosine in the biosynthesis of proteins such as β-galactosidases (Escherichia coli), bacteriorhodopsin and intestinal microvillar enzymes, aminopeptidase N, to study the effect of halogenated tryosines on the proteins properties.
Solubility
Soluble in water (28 g/L).
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
403-90-7 | |
199.18 | |
VIIAUOZUUGXERI-UHFFFAOYNA-N | |
92100 | |
2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid |
C9H10FNO3 | |
MFCD00063075 | |
3-fluoro-dl-tyrosine, 2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, pardinon, m-fluoro-dl-tyrosine, fluorthyrin, 3-fluorotyrosin, 3-fluorotyrosine, m-ftyr, dl-m-fluorotyrosine, dl-3-fluorotyrosine | |
CHEBI:32771 | |
NC(CC1=CC=C(O)C(F)=C1)C(O)=O |
Specifications
∽280°C (decomposition) | |
C9H10FNO3 | |
3204804 | |
Soluble in water (28g/L). | |
NC(CC1=CC=C(O)C(F)=C1)C(O)=O | |
199.18 | |
CHEBI:32771 | |
98% | |
3-Fluoro-DL-tyrosine |
403-90-7 | |
MFCD00063075 | |
3-fluoro-dl-tyrosine, 2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, pardinon, m-fluoro-dl-tyrosine, fluorthyrin, 3-fluorotyrosin, 3-fluorotyrosine, m-ftyr, dl-m-fluorotyrosine, dl-3-fluorotyrosine | |
VIIAUOZUUGXERI-UHFFFAOYNA-N | |
2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid | |
92100 | |
199.18 | |
100 mg |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 206-964-0
RTECSNumber : YP2660000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only