missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-DL-phenylglycine, 97+%

$359.03 - $359.03

Chemical Identifiers

CAS 7292-74-2
Molecular Formula C8H8FNO2
Formula Weight 169.16
MDL Number MFCD00049325
Chemical Name or Material 3-Fluoro-DL-phenylglycine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2281209 SDS Alfa Aesar™
B2281209
10g Each for $359.03
AAB2281204 SDS Alfa Aesar™
B2281204
2g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
3-Fluoro-DL-phenylglycine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

7292-74-2
169.16
3-Fluoro-DL-phenylglycine
C8H8FNO2
MFCD00049325

Specifications

7292-74-2
169.16
≈290°C (subl.)
3-Fluoro-DL-phenylglycine
C8H8FNO2
MFCD00049325
10g
≥97%