Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)phenol, 97+%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)phenol
CAS 172333-87-8
Molecular Formula C7H4F4O
Formula Weight 180.1
MDL Number MFCD00061288
UN Number UN3265
Synonym alpha,alpha,alpha,5-Tetrafluoro-m-cresol; 5-Fluoro-alpha,alpha,alpha-trifluoro-m-cresol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2456504

  SDS
alfa aesar™
B2456504
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2456509

  SDS
alfa aesar™
B2456509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 180.1
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN3265
Health Hazard 2 GHS H Statement
H301-H314-H318-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Molecular Formula C7H4F4O
MDL Number MFCD00061288
Health Hazard 1 Danger
Quantity 2g
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Assay ≥97%
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)phenol
Synonym alpha,alpha,alpha,5-Tetrafluoro-m-cresol; 5-Fluoro-alpha,alpha,alpha-trifluoro-m-cresol
CAS 172333-87-8
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.