missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)benzylamine, 97%

Manufacturer:  Alfa Aesar™ L1083603

 View more versions of this product

Catalog No. AAL1083603



Description

Description

Applications
3-Fluoro-5-(trifluoromethyl)benzylamine may be employed as amine to generate different trityl-based pro-tags, by treating with activated carboxyl group-containing trityl synthons.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Specifications

Specifications

3-Fluoro-5-(trifluoromethyl)benzylamine
1g
C8H7F4N
UN2735
3-fluoro-5-trifluoromethyl benzylamine, 3-fluoro-5-trifluoromethyl phenyl methanamine, 3-fluoro-5-trifluoromethylbenzylamine, 3-fluoro-5-trifluoromethyl phenyl methanamine, 3-fluoro-5-trifluoromethyl-benzylamine, benzenemethanamine, 3-fluoro-5-trifluoromethyl, 1-3-fluoro-5-trifluoromethyl phenyl methanamine, 5-fluoro-3-trifluoromethyl phenyl methylamine, acmc-1c7bq, ksc495o0t
C1=C(C=C(C=C1C(F)(F)F)F)CN
193.145
193.15
76°C (168°F)
150517-77-4
MFCD00061163
7807166
ADHUUMWSWNPEMZ-UHFFFAOYSA-N
[3-fluoro-5-(trifluoromethyl)phenyl]methanamine
2733304
97%

Chemical Identifiers

150517-77-4
193.145
ADHUUMWSWNPEMZ-UHFFFAOYSA-N
2733304
C1=C(C=C(C=C1C(F)(F)F)F)CN
C8H7F4N
MFCD00061163
3-fluoro-5-trifluoromethyl benzylamine, 3-fluoro-5-trifluoromethyl phenyl methanamine, 3-fluoro-5-trifluoromethylbenzylamine, 3-fluoro-5-trifluoromethyl phenyl methanamine, 3-fluoro-5-trifluoromethyl-benzylamine, benzenemethanamine, 3-fluoro-5-trifluoromethyl, 1-3-fluoro-5-trifluoromethyl phenyl methanamine, 5-fluoro-3-trifluoromethyl phenyl methylamine, acmc-1c7bq, ksc495o0t
[3-fluoro-5-(trifluoromethyl)phenyl]methanamine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

Documents