Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)benzyl alcohol, 97%

$89.73 - $89.73

Chemical Identifiers

Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzyl alcohol
CAS 184970-29-4
Molecular Formula C8H6F4O
Formula Weight 194.13
MDL Number MFCD00061169
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2038703

 
alfa aesar™
B2038703
1g Each for $89.73
Description & Specifications

Specifications

Formula Weight 194.13
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzyl alcohol
Assay 97%
Molecular Formula C8H6F4O
MDL Number MFCD00061169
CAS 184970-29-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 80° to 82°C (3mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.