Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)benzoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzoyl chloride
CAS 171243-30-4
Molecular Formula C8H3ClF4O
Formula Weight 226.56
MDL Number MFCD00061157
UN Number UN3265
Beilstein 8056839
Synonym alpha,alpha,alpha,5-Tetrafluoro-m-toluoyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2049903

 
alfa aesar™
B2049903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2049906

 
alfa aesar™
B2049906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 226.56
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzoyl chloride
MDL Number MFCD00061157
UN Number UN3265
Flash Point 73°C (163°F)
Quantity 1g
Assay 97%
Molecular Formula C8H3ClF4O
CAS 171243-30-4
Beilstein 8056839
Synonym alpha,alpha,alpha,5-Tetrafluoro-m-toluoyl chloride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Boiling Point ≈170°C
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.