missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoro-5-(trifluoromethyl)benzoic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442600010

 View more versions of this product

Catalog No. AC442600010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Fluoro-5-(trifluoromethyl)benzoic acid
105.0°C to 106.0°C
161622-05-5
C8H4F4O2
NSGKIIGVPBTOBF-UHFFFAOYSA-N
3-fluoro-5-(trifluoromethyl)benzoic acid
519222
98%
Glass Bottle
1g
97.5% min. (HPLC)
3-fluoro-5-trifluoromethyl benzoic acid, 5-fluoro-3-trifluoromethyl benzoic acid, acmc-1bwlq, alpha,alpha,alpha,5-tetrafluoro-m-toluic acid, benzoic acid, 3-fluoro-5-trifluoromethyl, buttpark 32\01-71, intermediates-zcf02214, ksc496a7r, maybridge4_001933, rarechem al bo 0634
C1=C(C=C(C=C1C(F)(F)F)F)C(=O)O
208.11
208.11

Chemical Identifiers

161622-05-5
208.11
3-fluoro-5-trifluoromethyl benzoic acid, 5-fluoro-3-trifluoromethyl benzoic acid, acmc-1bwlq, alpha,alpha,alpha,5-tetrafluoro-m-toluic acid, benzoic acid, 3-fluoro-5-trifluoromethyl, buttpark 32\01-71, intermediates-zcf02214, ksc496a7r, maybridge4_001933, rarechem al bo 0634
3-fluoro-5-(trifluoromethyl)benzoic acid
C8H4F4O2
NSGKIIGVPBTOBF-UHFFFAOYSA-N
519222
C1=C(C=C(C=C1C(F)(F)F)F)C(=O)O
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS Documents