missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Fluoro-5-(trifluoromethyl)benzoic acid, 98%, ACROS Organics™

Chemical Identifiers

CAS 161622-05-5
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.11
InChI Key NSGKIIGVPBTOBF-UHFFFAOYSA-N
Synonym 3-fluoro-5-trifluoromethyl benzoic acid, 5-fluoro-3-trifluoromethyl benzoic acid, acmc-1bwlq, alpha,alpha,alpha,5-tetrafluoro-m-toluic acid, benzoic acid, 3-fluoro-5-trifluoromethyl, buttpark 32\01-71, intermediates-zcf02214, ksc496a7r, maybridge4_001933, rarechem al bo 0634
PubChem CID 519222
IUPAC Name 3-fluoro-5-(trifluoromethyl)benzoic acid
SMILES C1=C(C=C(C=C1C(F)(F)F)F)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442600010
SDS View Documents
Thermo Fisher Scientific
AC442600010
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC442600050
SDS View Documents
Thermo Fisher Scientific
AC442600050
5g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

161622-05-5
208.11
3-fluoro-5-trifluoromethyl benzoic acid, 5-fluoro-3-trifluoromethyl benzoic acid, acmc-1bwlq, alpha,alpha,alpha,5-tetrafluoro-m-toluic acid, benzoic acid, 3-fluoro-5-trifluoromethyl, buttpark 32\01-71, intermediates-zcf02214, ksc496a7r, maybridge4_001933, rarechem al bo 0634
3-fluoro-5-(trifluoromethyl)benzoic acid
C8H4F4O2
NSGKIIGVPBTOBF-UHFFFAOYSA-N
519222
C1=C(C=C(C=C1C(F)(F)F)F)C(=O)O

Specifications

Glass Bottle
1g
97.5% min. (HPLC)
3-fluoro-5-trifluoromethyl benzoic acid, 5-fluoro-3-trifluoromethyl benzoic acid, acmc-1bwlq, alpha,alpha,alpha,5-tetrafluoro-m-toluic acid, benzoic acid, 3-fluoro-5-trifluoromethyl, buttpark 32\01-71, intermediates-zcf02214, ksc496a7r, maybridge4_001933, rarechem al bo 0634
C1=C(C=C(C=C1C(F)(F)F)F)C(=O)O
208.11
208.11
3-Fluoro-5-(trifluoromethyl)benzoic acid
105.0°C to 106.0°C
161622-05-5
C8H4F4O2
NSGKIIGVPBTOBF-UHFFFAOYSA-N
3-fluoro-5-(trifluoromethyl)benzoic acid
519222
98%
Safety and Handling

Safety and Handling

GHS Signal Word: Warning