Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)benzoic acid, 98%

$59.38 - $239.58

Chemical Identifiers

Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzoic acid
CAS 161622-05-5
Molecular Formula C8H4F4O2
Formula Weight 208.11
MDL Number MFCD00061293
Synonym alpha,alpha,alpha,5-Tetrafluoro-m-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1851603

  SDS
alfa aesar™
A1851603
1g Each for $59.38

AAA1851606

  SDS
alfa aesar™
A1851606
5g Each for $239.58
Description & Specifications

Specifications

Formula Weight 208.11
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
Health Hazard 1 Warning
Melting Point 103° to 105°C
Molecular Formula C8H4F4O2
MDL Number MFCD00061293
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym alpha,alpha,alpha,5-Tetrafluoro-m-toluic acid
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzoic acid
Assay 98%
CAS 161622-05-5
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.