Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)benzamide, 97%

$88.59 - $305.88

Chemical Identifiers

Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzamide
CAS 207986-20-7
Molecular Formula C8H5F4NO
Formula Weight 207.13
MDL Number MFCD00061149
Synonym alpha,alpha,alpha,5-Tetrafluoro-m-toluamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1285403

 
alfa aesar™
L1285403
1g Each for $88.59

AAL1285406

 
alfa aesar™
L1285406
5g Each for $305.88
Description & Specifications

Specifications

Formula Weight 207.13
Chemical Name or Material 3-Fluoro-5-(trifluoromethyl)benzamide
Molecular Formula C8H5F4NO
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
CAS 207986-20-7
MDL Number MFCD00061149
Assay 97%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym alpha,alpha,alpha,5-Tetrafluoro-m-toluamide
Melting Point 115° to 117°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.