missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Fluoro-5-(trifluoromethyl)benzamide, 97%

Manufacturer:  Alfa Aesar™ L1285406

 View more versions of this product

Catalog No. AAL1285406



Specifications

Specifications

3-Fluoro-5-(trifluoromethyl)benzamide
5g
C8H5F4NO
3-fluoro-5-trifluoromethyl benzamide, 5-fluoro-3-trifluoromethyl benzamide, acmc-1cmgv, 3-fluoro-5-trifluoromethyl-benzamide, 3-fluoranyl-5-trifluoromethyl benzamide, benzamide,3-fluoro-5-trifluoromethyl
C1=C(C=C(C=C1C(F)(F)F)F)C(=O)N
207.128
207.13
115°C to 117°C
207986-20-7
MFCD00061149
BBUBHNZCRQRAQG-UHFFFAOYSA-N
3-fluoro-5-(trifluoromethyl)benzamide
2737544
97%

Chemical Identifiers

207986-20-7
207.128
BBUBHNZCRQRAQG-UHFFFAOYSA-N
2737544
C1=C(C=C(C=C1C(F)(F)F)F)C(=O)N
C8H5F4NO
MFCD00061149
3-fluoro-5-trifluoromethyl benzamide, 5-fluoro-3-trifluoromethyl benzamide, acmc-1cmgv, 3-fluoro-5-trifluoromethyl-benzamide, 3-fluoranyl-5-trifluoromethyl benzamide, benzamide,3-fluoro-5-trifluoromethyl
3-fluoro-5-(trifluoromethyl)benzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents